missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethylaminocarbonyl)-benzolboronsäurepinakolester, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030329
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-60-2
C16H24BNO4
305.18
MFCD09266188
XNDAIQVEKVZONM-UHFFFAOYSA-N
n-2-methoxyethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-methoxyethylcarbamoyl phenylboronic acid, pinacol ester, 4-2-methoxyethylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb042, 4-2-methoxyethyl carbamoyl phenylboronic acid pinacol ester, 4-2-methoxyethylaminecarbonyl benzeneboronic acid pinacol ester, n-2-methoxyethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739044
N-(2-Methoxyethyl)-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Benzamid
COCCNC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-60-2
C16H24BNO4
305.18
MFCD09266188
XNDAIQVEKVZONM-UHFFFAOYSA-N
n-2-methoxyethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-methoxyethylcarbamoyl phenylboronic acid, pinacol ester, 4-2-methoxyethylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb042, 4-2-methoxyethyl carbamoyl phenylboronic acid pinacol ester, 4-2-methoxyethylaminecarbonyl benzeneboronic acid pinacol ester, n-2-methoxyethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739044
N-(2-Methoxyethyl)-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Benzamid
COCCNC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
Schmelzpunkt 73°C to 76°C
Menge 1 g
Formelmasse 305.18
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'