Learn More
4-(6-Thiocarbamoyl-2-benzimidazolyl)-thiobenzamid, 97 %, Thermo Scientific™
Catalog Number 15449625
Click to view available options
Menge:
250 mg
1 g
1256482-81-1
C15H12N4S2
312.41
MFCD16658839
YFZMXNBFKPYLTB-UHFFFAOYSA-N
4-6-thiocarbamoyl-2-benzimidazolyl thiobenzamide, 2-4-carbamothioylphenyl-3h-1,3-benzodiazole-5-carbothioamide
73995300
2-(4-Carbamothioylphenyl)-3H-Benzimidazol-5-Carbothioamid
NC(=S)C1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C(N)=S
1256482-81-1
C15H12N4S2
312.41
MFCD16658839
YFZMXNBFKPYLTB-UHFFFAOYSA-N
4-6-thiocarbamoyl-2-benzimidazolyl thiobenzamide, 2-4-carbamothioylphenyl-3h-1,3-benzodiazole-5-carbothioamide
73995300
2-(4-Carbamothioylphenyl)-3H-Benzimidazol-5-Carbothioamid
NC(=S)C1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C(N)=S
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(6-Thiocarbamoyl-2-benzimidazolyl)thiobenzamide
Menge 250 mg
Formelmasse 312.41
Reinheit (%) 97%