Learn More
DiscoverX 4-CMTB
Artikelnummer. 17392609
Encompass
Ansicht ändern
Click to view available options
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17392609 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17392609 Lieferant DiscoverX Lieferanten-Nr. 921300
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
4-CMTB is an agonist for FFA2. Provided as 10mg of dry powder.
4-CMTB or 4-chloro-α-(1-methylethyl)-N-2-thiazolylbenzeneacetamide, is a free fatty acid receptor FFA2 agonist and positive allosteric modulator. Binds at a site distinct from the orthosteric site; modulates the activity of short-chain fatty acids at FFA2 via the FFA2 second extracellular loop (ECL2). Provided as 10mg of dry powder.
Spezifikation
Menge 10 mg