missing translation for 'onlineSavingsMsg'
Learn More

4-Fluor-3-(4-methylphenylcarbamoyl)benzenboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7036562
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096333-11-6
C14H13BFNO3
273.07
MFCD20040261
ZBHFKQPACRWNAG-UHFFFAOYSA-N
4-fluoro-3-4-methylphenylcarbamoyl benzeneboronic acid
73996007
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096333-11-6
C14H13BFNO3
273.07
MFCD20040261
ZBHFKQPACRWNAG-UHFFFAOYSA-N
4-fluoro-3-4-methylphenylcarbamoyl benzeneboronic acid
73996007

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Fluoro-3-(4-methylphenylcarbamoyl)benzeneboronic acid
Menge 250 mg
Formelmasse 273.07
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'