missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)-benzolboronsäurepinakolester, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030469
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
171364-80-0
C14H19BO4
262.11
MFCD02179438
REIZEQZILPXYKS-UHFFFAOYSA-N
methyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonylphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid pinacolate, 4-carbomethoxyphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid, pinacol ester, 4-methoxycarbonyl phenylboronic acid pinacol ester, methyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, 4-methoxycarbonyl benzeneboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid methyl ester
2773500
Methyl 4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)benzoat
COC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
171364-80-0
C14H19BO4
262.11
MFCD02179438
REIZEQZILPXYKS-UHFFFAOYSA-N
methyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonylphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid pinacolate, 4-carbomethoxyphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid, pinacol ester, 4-methoxycarbonyl phenylboronic acid pinacol ester, methyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, 4-methoxycarbonyl benzeneboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid methyl ester
2773500
Methyl 4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)benzoat
COC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester
Schmelzpunkt 77°C to 81°C
Menge 1 g
Formelmasse 262.11
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'