missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Chlor-6-Nitrophenylcarbamoyl)-2-Fluorbenzenboronsäure, 97 %, Thermo Scientific™

Catalog Number 15471526
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096342-07-1
C13H9BClFN2O5
338.48
MFCD20126194
XRVHUHAJXXHBAJ-UHFFFAOYSA-N
5-2-chloro-6-nitrophenylcarbamoyl-2-fluorobenzeneboronic acid
73995938
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096342-07-1
C13H9BClFN2O5
338.48
MFCD20126194
XRVHUHAJXXHBAJ-UHFFFAOYSA-N
5-2-chloro-6-nitrophenylcarbamoyl-2-fluorobenzeneboronic acid
73995938

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-(2-Chloro-6-nitrophenylcarbamoyl)-2-fluorobenzeneboronic acid
Menge 250 mg
Formelmasse 338.49
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'