missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dichlor-1H-benzo[d]imidazol-2-thiol, ≥ 95 %, Thermo Scientific™
Click to view available options
Menge:
1 g
5 g
Specifications
Chemischer Name oder Material | 5, 6-Dichloro-1H-benzo[d]imidazole-2-thiol |
CAS Min. % | 94.5 |
CAS Max. % | 100.0 |
Prozentgehaltsbereich | 95% |
Verpackung | Bernsteinfarbene Glasflasche |
Menge | 1 g |
Formelmasse | 219.09 |
Reinheit (%) | 95% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'