missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress 5(6)-TAMRA SE
Specifications
Specifications
| Menge | 100 mg |
| Produkttyp | Dye Reagents |
| Molekulargewicht | 527.53 g/mol |
| Löslichkeit | DMSO : 10 mg/mL (18.96 mM; ultrasonic and warming and heat to 60°C) |
| Inhalt und Lagerung | -20°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen) |
| Versandbedingung | Room temperature |
| Ziel | HBV |
| CAS | 246256-50-8 |
| Synonym | 5(6)-Carboxytetramethylrhodamine N-succinimidyl ester |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?