missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4,6-triiodisophthalsäure, 95 %, Thermo Scientific Chemicals

Catalog Number p-7026263
Click to view available options
Menge:
10 g
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
35453-19-1
C8H4I3NO4
558.836
MFCD00190167
JEZJSNULLBSYHV-UHFFFAOYSA-N
5-amino-2,4,6-triiodoisophthalic acid, unii-a17g990rt0, 5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid, 5-amino-2,4,6-iodoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo, 5-amino-2,4,6-triiodoisophthalicacid, isophthalic acid, 5-amino-2,4,6-triiodo, acmc-209ih0, ksc224s6b, aronis25361
3015783
5-Amino-2,4,6-Triiodobenzol-1,3-Dicarbonsäure
C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
35453-19-1
C8H4I3NO4
558.836
MFCD00190167
JEZJSNULLBSYHV-UHFFFAOYSA-N
5-amino-2,4,6-triiodoisophthalic acid, unii-a17g990rt0, 5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid, 5-amino-2,4,6-iodoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo, 5-amino-2,4,6-triiodoisophthalicacid, isophthalic acid, 5-amino-2,4,6-triiodo, acmc-209ih0, ksc224s6b, aronis25361
3015783
5-Amino-2,4,6-Triiodobenzol-1,3-Dicarbonsäure
C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Amino-2,4,6-triiodoisophthalic acid
Schmelzpunkt 265°C to 270°C
Menge 10 g
Empfindlichkeit Light and moisture sensitive
Formelmasse 558.84
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'