missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4-methoxyphenyl)-1H-pyrazol, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030736
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
19541-95-8
C10H11N3O
189.218
MFCD00462192
UPAGEJODHNVJNM-UHFFFAOYSA-N
3-4-methoxyphenyl-1h-pyrazol-5-amine, 5-4-methoxyphenyl-1h-pyrazol-3-amine, 3-amino-5-4-methoxyphenyl pyrazole, 5-amino-3-4-methoxyphenyl pyrazole, 3-amino-5-4-methoxyphenyl-1h-pyrazole, 5-4-methoxy-phenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methoxyphenyl-1h-pyrazole, 1h-pyrazol-3-amine, 5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-pyrazol-3-amine, 5-4-methoxyphenyl-2h-pyrazol-3-ylamine
2756469
5-(4-Methoxyphenyl)-1H-Pyrazol-3-Amin
COC1=CC=C(C=C1)C2=CC(=NN2)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
19541-95-8
C10H11N3O
189.218
MFCD00462192
UPAGEJODHNVJNM-UHFFFAOYSA-N
3-4-methoxyphenyl-1h-pyrazol-5-amine, 5-4-methoxyphenyl-1h-pyrazol-3-amine, 3-amino-5-4-methoxyphenyl pyrazole, 5-amino-3-4-methoxyphenyl pyrazole, 3-amino-5-4-methoxyphenyl-1h-pyrazole, 5-4-methoxy-phenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methoxyphenyl-1h-pyrazole, 1h-pyrazol-3-amine, 5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-pyrazol-3-amine, 5-4-methoxyphenyl-2h-pyrazol-3-ylamine
2756469
5-(4-Methoxyphenyl)-1H-Pyrazol-3-Amin
COC1=CC=C(C=C1)C2=CC(=NN2)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Amino-3-(4-methoxyphenyl)-1H-pyrazole
Schmelzpunkt 141°C to 143°C
Menge 1 g
Formelmasse 189.22
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'