missing translation for 'onlineSavingsMsg'
Learn More

5-Butyldihydrofuran-2(3H)-one, 98%, BLD Pharm™

Artikelnummer. 31033079
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31033079 1 kg
31015157 25 g
31021437 100 g
31020436 500 g
31031560 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31033079 Lieferant BLD Pharm Lieferanten-Nr. BD1318691000g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 5-Butyldihydrofuran-2(3H)-one
CAS 104-50-7
Summenformel C8H14O2
MDL-Nummer MFCD00005402
Menge 1 kg
SMILES O=C1OC(CCCC)CC1
Molekulargewicht (g/mol) 142.20
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.