missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorimidazo-[1,2-a]-pyridin-2-carbonsäure Hydrat, 95 %, Thermo Scientific™

Catalog Number 15478975
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1000017-93-5
C8H5ClN2O2
196.59
MFCD09836168
TVUHAHMCORHJBN-UHFFFAOYSA-N
5-chloroimidazo 1,2-a pyridine-2-carboxylic acid, 5-chloro-imidazo 1,2-a pyridine-2-carboxylic acid, 5-chloroimidazo 1,2-a pyridine-2-carboxylic acid hydrate, 2-carboxy-5-chloroimidazo 1,2-a pyridine, 5-chloroimidazo 1,2-a pyridine-2-carboxylicacid, imidazo 1,2-a pyridine-2-carboxylicacid,5-chloro, imidazo 1,2-a pyridine-2-carboxylic acid, 5-chloro, 5-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid
26369183
5-Chlorimidazo[1,2-a]Pyridin-2-Carbonsäure
OC(=O)C1=CN2C(C=CC=C2Cl)=N1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1000017-93-5
C8H5ClN2O2
196.59
MFCD09836168
TVUHAHMCORHJBN-UHFFFAOYSA-N
5-chloroimidazo 1,2-a pyridine-2-carboxylic acid, 5-chloro-imidazo 1,2-a pyridine-2-carboxylic acid, 5-chloroimidazo 1,2-a pyridine-2-carboxylic acid hydrate, 2-carboxy-5-chloroimidazo 1,2-a pyridine, 5-chloroimidazo 1,2-a pyridine-2-carboxylicacid, imidazo 1,2-a pyridine-2-carboxylicacid,5-chloro, imidazo 1,2-a pyridine-2-carboxylic acid, 5-chloro, 5-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid
26369183
5-Chlorimidazo[1,2-a]Pyridin-2-Carbonsäure
OC(=O)C1=CN2C(C=CC=C2Cl)=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Schmelzpunkt 140°C to 142°C
Menge 1 g
Formelmasse 196.59 (Anhydrous)
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'