missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-phenyl-4-(2,2,2-trifluoroacetyl)-1H-pyrazol-3(2H)-one, 98%, BLD Pharm™

Product Code. 30787658
Encompass_Preferred
Change view
Click to view available options
Menge:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30787658 1 g
30786532 250 mg
30787103 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30787658 Supplier BLD Pharm Supplier No. BD144401g
 Eingeschränkter Artikel
Verfügbar ab: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 5-Methyl-2-phenyl-4-(2,2,2-trifluoroacetyl)-1H-pyrazol-3(2H)-one
CAS 1691-93-6
Summenformel C12H9F3N2O2
MDL-Nummer MFCD00143378
Menge 1 g
SMILES CC1=C(C(=O)C(F)(F)F)C(=O)N(N1)C1=CC=CC=C1
Molekulargewicht (g/mol) 270.21
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.