missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1,3-oxazole, 97%, BLD Pharm™

Product Code. 31033961
Encompass_Preferred
Change view
Click to view available options
Menge:
100 mg
250 mg
1 g
5 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31033961 100 mg
31021295 25 g
31016918 1 g
31030787 5 g
31024447 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31033961 Supplier BLD Pharm Supplier No. BD105267100mg
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 5-Phenyl-1,3-oxazole
CAS 1006-68-4
Summenformel C9H7NO
MDL-Nummer MFCD00067074
Menge 100 mg
SMILES O1C=NC=C1C1=CC=CC=C1
Molekulargewicht (g/mol) 145.16
Reinheit (%) 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.