Learn More
Pan Biotech 500ML Alpha MEM Eagle, w: L-Glutamine, w/o: Ribonucleosides, w/o: Deoxyribonucleosides, w: 2.2g/L Na
Marke:  Pan Biotech P04-21060
Encompass
500ML Alpha MEM Eagle, w: L-Glutamine, w/o: Ribonucleosides, w/o: Deoxyribonucleosides, w: 2.2g/L Na
Artikelnummer. 17120879
  • 35.70 € / 500 Milliliter