missing translation for 'onlineSavingsMsg'
Learn More

6-Brompyridin-2-boronsäurepinakolester, 95 %, Thermo Scientific Chemicals

Catalog Number 15467665
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
651358-83-7
C11H15BBrNO2
283.96
MFCD06796177
OAVRLCKBKDMGGW-UHFFFAOYSA-N
6-bromopyridine-2-boronic acid pinacol ester, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester
12177444
2-Brom-6-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Pyridin
B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
651358-83-7
C11H15BBrNO2
283.96
MFCD06796177
OAVRLCKBKDMGGW-UHFFFAOYSA-N
6-bromopyridine-2-boronic acid pinacol ester, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester
12177444
2-Brom-6-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Pyridin
B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Bromopyridine-2-boronic acid pinacol ester
Schmelzpunkt 125°C to 127°C
Menge 250 mg
Formelmasse 283.96
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'