missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxynicotinsäure, 98 %, Thermo Scientific Chemicals

Catalog Number p-7022851
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5006-66-6
C6H5NO3
139.11
MFCD00006277
BLHCMGRVFXRYRN-UHFFFAOYSA-N
6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid
72924
CHEBI:16168
6-Oxo-1H-Pyridin-3-Carbonsäure
C1=CC(=O)NC=C1C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5006-66-6
C6H5NO3
139.11
MFCD00006277
BLHCMGRVFXRYRN-UHFFFAOYSA-N
6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid
72924
CHEBI:16168
6-Oxo-1H-Pyridin-3-Carbonsäure
C1=CC(=O)NC=C1C(=O)O

Metabolite of nicotinamide. Stimulates adrenocortical secretion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Hydroxynicotinic acid
Schmelzpunkt 314°C to 316°C
Menge 5 g
Beilstein 472182
Formelmasse 139.11
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'