missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-methylpyridin-3-Boronsäure-Pinacolester, 96 %, Thermo Scientific™

Catalog Number 15461516
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1080028-73-4
C13H20BNO3
249.117
MFCD12923398
CWJKZNNXSPVPAY-UHFFFAOYSA-N
6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methylpyridine-3-boronic acid pinacol ester, \rtf1\ansi\deff0 \fonttbl \f0\fnil\fcharset0 ms sans serif;, 6-methoxy-2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-boronic acid pinacol ester pyridine, 6-methoxy-2-methyl-pyridine-3-boronic acid pinacol ester, 2-methyl-6-methoxypyridine-3-boronic acid pinacol ester, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine
67487482
6-Methoxy-2-Methyl-3-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)pyridin
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)OC)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1080028-73-4
C13H20BNO3
249.117
MFCD12923398
CWJKZNNXSPVPAY-UHFFFAOYSA-N
6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methylpyridine-3-boronic acid pinacol ester, \rtf1\ansi\deff0 \fonttbl \f0\fnil\fcharset0 ms sans serif;, 6-methoxy-2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-boronic acid pinacol ester pyridine, 6-methoxy-2-methyl-pyridine-3-boronic acid pinacol ester, 2-methyl-6-methoxypyridine-3-boronic acid pinacol ester, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine
67487482
6-Methoxy-2-Methyl-3-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)pyridin
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)OC)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester
Menge 1 g
Formelmasse 249.11
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'