Learn More
Ibidi 8 Well Chamber, Removable, Bulk Pack, 8 per tray, 12 trays
Beschreibung
- Chamber slide with a removable 8 well silicone chamber for cell culture and immunofluorescence staining
- for use with upright and inverted microscopes
- bottom material: microscopy glass
- packed in trays: 8 per tray
- sterilized
Spezifikation
Spezifikation
| Typ | Chambered Coverglass |
| Wells | 8 |
| Sterilität | Steril |