missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A20A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539025
Weitere Details : Netto-Gewicht : 0.00900kg
Spezifikation
A20A3 | |
Polyclonal | |
Unconjugated | |
ANKRD20A3 | |
ANKRD20A3 | |
A synthetic peptide derived from the internal region of human ANKRD20A3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5VUR7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441425 | |
-20°C | |
Liquid |