Learn More
ABAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527405
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: SK-N-SH, mouse brain, rat liver. Predicted reactivity: Mouse (93%), Rat (93%), Xenopus laevis (82%), Pig (95%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Spezifikation
ABAT | |
Polyclonal | |
Unconjugated | |
ABAT | |
(S)-3-amino-2-methylpropionate transaminase; 4-aminobutyrate aminotransferase; 4-aminobutyrate aminotransferase, brain isoform; 4-aminobutyrate aminotransferase, liver isoform; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; 9630038C02Rik; ABAT; AI255750; beta-AlaAT; beta-alanine oxoglutarate aminotransferase; cb880; ENSMUSG00000051226; fj82a01; FLJ17813; FLJ30272; GABA aminotransferase; GABA AT; GABA T; GABA transaminase; GABA transferase; Gabaat; GABA-AT; Gabat; GABA-T; Gamma-amino-N-butyrate transaminase; Gm9851; hCG1984265; I54; L AIBAT; Laibat; L-AIBAT; mitochondrial 4-aminobutyrate aminotransferase; NPD009; wu:fj82a01; X61497 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18, 268860, 81632 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.49 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P50554, P61922, P80404 | |
ABAT | |
Recombinant fragment corresponding to a region within amino acids 302 and 500 of Human ABAT | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |