missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA1 Monoclonal Antibody (AB1.G6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA191401
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-91401 detects ABCA1 from human samples.
ABCA1 Monoclonal antibody specifically detects ABCA1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Spezifikation
ABCA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.4 | |
O95477, P41233 | |
Abca1 | |
amino acids 1800-2260 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
AB1.G6 | |
Unconjugated | |
Abca1 | |
ABC 1; ABC 1 antibody; ABC Transporter 1; Abc1; ABC-1; ABC1 antibody; Abca1; ATP binding cassette subfamily A member 1; ATP-binding cassette 1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, subfamily A (ABC1), member 1; ATP-binding cassette, sub-family A (ABC1), member 1; ATP-binding cassette, sub-family A member 1; CERP; CERP antibody; Cholesterol efflux regulatory protein; HDLDT1; membrane-bound; phospholipid-transporting ATPase ABCA1; phospholipid-transporting ATPase ABCA1; ATP-binding cassette sub-family A member 1; TGD | |
Mouse | |
IgG fraction | |
RUO | |
11303, 19, 313210 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |