Learn More
ABCA10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553186
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MYKVTRETHC WEFSPSMYFL SLEQIPKTPL TSLLIVNNTG SNIEDLVHSL KCQDIVLEID DFRNRNGSDD PSYNGAIIVS GDQKDYRFSV ACNTKKLNC Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 60%.
ABCA10 (ATP Binding Cassette Subfamily A Member 10) is a Protein Coding gene. Diseases associated with ABCA10 include Donnai-Barrow Syndrome. Among its related pathways are Regulation of activated PAK-2p34 by proteasome mediated degradation and Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds. Gene Ontology (GO) annotations related to this gene include ATPase activity and ATPase activity, coupled to transmembrane movement of substances. An important paralog of this gene is ABCA9.Spezifikation
ABCA10 | |
Polyclonal | |
Unconjugated | |
ABCA10 | |
ABCA10; ATP binding cassette subfamily A member 10; ATP-binding cassette A10; ATP-binding cassette sub-family A member 10; ATP-binding cassette, sub-family A (ABC1), member 10; EST698739; FKHL6; FREAC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWZ4 | |
ABCA10 | |
Recombinant protein corresponding to Human ABCA10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only