missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA549985
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ABCA13 protein.
In human, the ATP-binding cassette (ABC) family of transmembrane transporters has at least 48 genes and 7 gene subfamilies. This gene is a member of ABC gene subfamily A (ABCA). Genes within the ABCA family typically encode several thousand amino acids. Like other ABC transmembrane transporter proteins, this protein has 12 or more transmembrane alpha-helix domains that likely arrange to form a single central chamber with multiple substrate binding sites. It is also predicted to have two large extracellular domains and two nucleotide binding domains as is typical for ABCA proteins. Alternative splice variants have been described but their biological validity has not been demonstrated.[provided by RefSeq, Mar 2009].Spezifikation
ABCA13 | |
Polyclonal | |
Unconjugated | |
ABCA13 | |
ABCA13; ATP binding cassette subfamily A member 13; ATP binding cassette transporter A13; ATP-binding cassette sub-family A member 13; ATP-binding cassette sub-family A member 13 variant 2; ATP-binding cassette sub-family A member 13 variant 3; ATP-binding cassette, sub-family A (ABC1), member 13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
154664 | |
-20°C | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q86UQ4 | |
ABCA13 | |
Synthesized peptide derived from internal of human ABCA13. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |