missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534396
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is 293 cell lysate. PA5-34396 can be used with blocking peptide PEP-1438.
ATP-binding cassette (ABC) transporters are an evolutionarily conserved family that use ATP hydrolysis to catalyze the transport of various molecules across cell membranes. They are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). ABCA7, a member of the ABC1 subfamily, is a full-size ABC transporter consisting of two sets of the multiple membrane-spanning domains plus the Walker motifs for the ATP interaction. ABCA7 shows the highest homology to ABCA1, an essential molecule for cholesterol homeostasis. The high expression levels of ABCA7 in peripheral leukocytes, thymus, spleen and bone marrow suggests a role in the immune system lipid homeostasis.Spezifikation
ABCA7 | |
Polyclonal | |
Unconjugated | |
Abca7 | |
ABC 1 antibody; ABC Transporter 1; ABC1; ABC1 antibody; Abc51; Abca7; ABCA-SSN; ABCX; AD9; ATP binding cassette subfamily A member 7; ATP-binding cassette sub-family A member 7; ATP-binding cassette sub-family A member 7; LOW QUALITY PROTEIN: ATP-binding cassette sub-family A member 7; ATP-binding cassette, subfamily A (ABC1), member 7; ATP-binding cassette, sub-family A (ABC1), member 7; ATP-binding cassette, sub-family A, member 7; autoantigen SS-N; CERP; CERP antibody; EC 2.2.1.1; EC 3.6.3; EC 3.6.3.41; FLJ40025; Macrophage ABC transporter; Phospholipid-transporting ATPase ABCA7; phospholipid-transporting ATPase ABCA7; ATP-binding cassette sub-family A member 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10347 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8IZY2 | |
Abca7 | |
An 18 amino acid synthetic peptide near the amino terminus of human ABCA7 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |