Learn More
ABCB11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553899
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RLSTVRAADT IIGFEHGTAV ERGTHEELLE RKGVYFTLVT LQSQGNQALN EEDIKDATED DMLARTFSRG SYQDSLRASI RQRSKSQLSY LVHEPPLAVV DHKSTYEEDR KDKDIPVQEE VEPAPVRRIL K Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
ABCB11 is involved in the ATP-dependent secretion of bile salts into the canaliculus of hepatocytes. It is expressed predominatly, if not exclusively, in the liver, where it is further localized to the canilicular microvilli and to subcanilicular vesicles fo the hepatocytes. Structurally, ABCB11 is a multifunctional polypeptide with two homologus halves, each containing a hydrophobic membrane-anchoring domain and an ATP binding cassette (ABC) domain. Defects in ABCB11 are the cause of progressive familial intrahepatic cholestasis 2 (PFIC2). PFIC2 is an inherited liver disease of childhood which is characterized by cholestasis and normal serum gamma-glutamyltransferase activity. Defects in ABCB11 are also found in cases of chronic intrahepatic cholestasis without obvious familial history of chronic liver disease.Spezifikation
ABCB11 | |
Polyclonal | |
Unconjugated | |
ABCB11 | |
ABC member 16, MDR/TAP subfamily; ABC16; Abcb11; ATP binding cassette subfamily B member 11; ATP-binding cassette sub-family B member 11; ATP-binding cassette, subfamily B (MDR/TAP), member 11; ATP-binding cassette, sub-family B (MDR/TAP), member 11; ATP-binding cassette, sub-family B, member 11; bile salt export pump; BRIC2; Bsep; Lith1; PFIC2; PFIC-2; PGY4; progressive familial intrahepatic cholestasis 2; sister of P-glycoprotein; sister p-glycoprotein; Spgp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8647 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.48 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95342 | |
ABCB11 | |
Recombinant protein corresponding to Human ABCB11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only