missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCF3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550288
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ABCF3 protein.
This protein may display an antiviral effect against flaviviruses such as west Nile virus (WNV) in the presence of OAS1B. ATP-binding cassette (ABC) transporters regulate the transport of a variety of physiologic substrates. But this protein lacks transmembrane domains and is probably not involved in transport.Spezifikation
ABCF3 | |
Polyclonal | |
Unconjugated | |
ABCF3 | |
ABCF3; AI326318; ATP binding cassette subfamily F member 3; ATP-binding cassette sub-family F member 3; ATP-binding cassette, subfamily F (GCN20), member 3; ATP-binding cassette, sub-family F (GCN20), member 3; AU016058; BB119416; EST201864 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55324 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NUQ8 | |
ABCF3 | |
Fusion protein corresponding to a region derived from internal residues of human ATP-binding cassette, sub-family F (GCN20), member 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |