missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26091-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is a member of the AB hydrolase superfamily and encodes a protein with an alpha/beta hydrolase fold. This domain is common to a number of hydrolytic enzymes of widely differing phylogenetic origins and catalytic functions.Spezifikation
ABHD1 | |
Polyclonal | |
Unconjugated | |
ABHD1 | |
ABHD1, LABH1, Lung alpha/beta hydrolase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57742, 84696 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96SE0, Q9QZC8 | |
ABHD1 | |
ABHD1 Fusion Protein Ag23138 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |