missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD17C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561831
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LDAVEVFFSR TARDNRLGCM FV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
ABHD17C Polyclonal antibody specifically detects ABHD17C in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ABHD17C | |
Polyclonal | |
Unconjugated | |
ABHD17C | |
2210412D01Rik; ABHD17C; abhydrolase domain containing 17C; abhydrolase domain-containing protein 17C; abhydrolase domain-containing protein FAM108C1; AL023007; alpha/beta hydrolase domain-containing protein 17C; Fam108c; Fam108c1; family with sequence similarity 108, member C; family with sequence similarity 108, member C1; protein ABHD17C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58489 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PCB6 | |
ABHD17C | |
Recombinant protein corresponding to Human ABHD17C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only