missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ Zubehör
Verzahnen Sie Ihr SymLine System genau nach Ihren Bedürfnissen! Hier finden Sie viele nützliche Ergänzungen.
Marke: SymLine™ 106605
Weitere Details : Netto-Gewicht : 1.60000kg
Beschreibung
- SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
- All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Spezifikation
Wall Mount | |
182 x 48 x 104 mm | |
SymLine™ System Customization |
White | |
Fire Protection Package, Mounting Kit, 28 mm Dia. Pipe Duct | |
Asecos™ Type 90 Safety Cabinet |