missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2AK5ac Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540102
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
H2AK5ac Polyclonal antibody specifically detects H2AK5ac in Human samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Peptide Array, Western Blot.Spezifikation
H2AK5ac | |
Polyclonal | |
Unconjugated | |
H2AX | |
1190022L06; Acetyl-Histone H2; Acetyl-Histone H2A K5; Acetyl-Histone H2A Lys5; AW228881; dJ160A22.4; dJ86C11.1; gammaH2ax; H2A 615; H2A GL101; H2A histone family member; H2A histone family member X; H2A histone family member Z; H2A histone family, member A; H2A histone family, member C; H2A histone family, member E; H2A histone family, member G; H2A histone family, member M; H2A histone family, member P; H2A histone family, member X; H2A histone family, member Z; H2A.1; H2A.1b; H2A.2; H2A.3; H2A.X; H2A.z; H2a.z-1; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/m; H2A/p; H2A/S; H2a/x; H2A/z; H2a-291A; H2a-53; H2ac11; H2ac13; H2AC15; H2AC16; H2AC17; h2ad; H2ae; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFM; H2AFN; H2AFP; H2afx; H2afz; H2AG; H2AH; h2ai; H2ak; H2AK5ac; H2ALys5ac; H2ax; H2AX histone; H2AZ; H2AZ histone; HIS; Hist1h2aa; HIST1H2AB; Hist1h2ac; HIST1H2AD; Hist1h2ae; HIST1H2AG; HIST1H2AH; Hist1h2ai; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; Hist1h2an; Hist1h2ao; Hist1h2ap; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221613, 255626, 3012, 3013, 3014, 3015, 8329, 8331, 8335, 85235, 8969 | |
-20°C or -80°C if preferred | |
Liquid |
ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Peptide Array, Western Blot | |
0.93 mg/mL | |
PBS with 0.05% sodium azide; pH 7.4 | |
P04908, P0C0S5, P0C0S8, P16104, P20671, Q96A08, Q96KK5, Q96QV6, Q99878 | |
H2ac1, H2AC11, H2AC12, H2AC13, H2AC14, H2AC4, H2AC7, H2AC8, H2AX, H2AZ1, H2bc1 | |
KLH-conjugated synthetic peptid corresponding to human the region of histone H2A containing the acetylated lysine 5 (H2AK5ac). | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |