Learn More
ACN9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554227
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YATALLQQAN ENRQNSTGKA CFGTFLPEEK LNDFRDEQIG QLQELMQEAT KPNRQFSISE SMKP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.
ACN9 Polyclonal antibody specifically detects ACN9 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
ACN9 | |
Polyclonal | |
Unconjugated | |
SDHAF3 | |
0610005M07Rik; 4933430A16Rik; Acn9; ACN9 homolog; ACN9 homolog (S. cerevisiae); DC11; Liver regeneration-related protein LRRGT00092; LRRGT00092; LYRM10; protein ACN9 homolog, mitochondrial; SDH assembly factor 3; Sdh7; SDHAF3; Succinate dehydrogenase assembly factor 3, mitochondrial; succinate dehydrogenase complex assembly factor 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57001 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRP4 | |
SDHAF3 | |
Recombinant protein corresponding to Human ACN9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only