Learn More
STEMCELL Technologies AggreWell™800 24-well Plate Starter Kit
Click to view available options
Anzahl Wells:
24
Packungsgröße:
1 Stück
Beschreibung
- AggreWell™ plates bring an easy, standardized approach to the generation of cell aggregates, including embryoid bodies (EBs) and spheroids
- EBs and spheroids generated using AggreWell™ plates are consistent in size and shape, and are uniform within and between experiments
- New and improved second-generation AggreWell™ plates are compatible with a variety of cell types, including ES and iPS cells, cancer cells and more
- Enhanced optical characteristics provide crystal clear imaging
- Note: AggreWell™ Rinsing Solution is required for optimal EB and spheroid formation
Spezifikation
Spezifikation
| Anzahl Wells | 24 |
| Zur Verwendung mit (Anwendung) | Differentiation; Spheroid Culture; Toxicity Assay |
| Menge | Starter Kit |