Learn More
AGO2 Mouse anti-Human, Unconjugated, Clone: 1G2H12, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 67934-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The AGO2 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human AGO2. This antibody recognizes human, mouse, rat antigen. The AGO2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
| AGO2 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 27161 | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 1G2H12 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |