missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1 Antitrypsin Monoclonal Antibody (TMF1#2-1D1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MIA1102
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
MIA1102 was formerly sold as a Seradyn product.
alpha-1 Antitrypsin Monoclonal antibody specifically detects alpha-1 Antitrypsin in Human samples. It is validated for ELISA, Flow Cytometry.Spezifikation
alpha-1 Antitrypsin | |
Monoclonal | |
2.76 mg/mL | |
PBS with 0.1% sodium azide | |
P01009 | |
SERPINA1 | |
Purified human alpha-1-antitrypsin | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry | |
TMF1#2-1D1 | |
Unconjugated | |
SERPINA1 | |
a-1 AT; A1A; a1-antitrypsin; A1AT; AAT; Aat2; Aat-2; alpha 1 antitrypsin; alpha-1 antitrypsin; alpha-1 protease inhibitor; Alpha-1 protease inhibitor 1; alpha-1-antiproteinase; Alpha-1-antitrypsin; alpha-1-antitrypsin (protease inhibitor); alpha-1-antitrypsin 1-1; alpha-1-antitrypsin 2; alpha-1-antitrypsin null; alpha1AT; alpha-1-protease inhibitor; alpha-1-proteinase inhibitor; Digestive Zymogen; Dom1; MGC23330; MGC9222; PI; PI1; PRO0684; PRO2209; PRO2275; protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin; protease inhibitor 1 (anti-elastase, antitrypsin, alpha-1); Protease Serine 1; serine (or cysteine) peptidase inhibitor, clade A, member 1A; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A (alpha-1antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1a; Serine Proteas | |
Mouse | |
Protein A | |
RUO | |
5265 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |