Learn More
Alpha 1B-Glycoprotein Mouse anti-Human, CoraLite 488, Clone: 4D4E5, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66260-100UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Alpha-1B-glycoprotein (A1BG) is a plasma protein and is a member of the immunoglobulin superfamily. A1BG contains 474 amino acids and has 5 intrachain disulfide bonds. It has been reported that cysteine-rich secretory protein 3 (CRISP-3) is a specific and high-affinity ligand of A1BG. A1BG-CRISP-3 complex displays a similar function in protecting the circulation from a potentially harmful effect of free CRISP3 (PMID: 15461460).
Spezifikation
Alpha 1B-Glycoprotein | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG1 |
Immunofluorescence | |
4D4E5 | |
CoraLite 488 | |
100 μL | |
1 | |
Liquid |