missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody (2A7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Marke: Novus Biologicals NBP1-05194
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Spezifikation
| alpha-Synuclein | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:1000, Simple Western 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000 | |
| P37840 | |
| SNCA | |
| This alpha-Synuclein Antibody (2A7) was developed against full length human recombinant protein with the epitope from amino acids 61-95. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Ascites |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| 2A7 | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| Mouse | |
| 15 kDa | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur