Learn More
Ansell™ AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped G01
Artikelnummer. p-200004960
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
4 Stück
Artikelnummer. 16884990
Marke: Ansell™ YE30T00075602G01
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
AlphaTec™ 3000 AVANT2 AIRline Ultrasonically welded and taped G01 is an encapsulated suit designed for use in combination with the permanently attached AlphaTec™ and AVANT STS V.1 airline regulator.
Encapsulated suit designed to be used with the permanently attached AlphaTec™ and AVANT STS V.1 airline regulator.
  • Includes internal air-channel ventilation system 
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
TRUSTED_SUSTAINABILITY
Specifications
Produkttyp Chemikalienschutzkleidung
Größe Small
Geschlecht Unisex
Material Multi-layer Barrier Laminate Fabric
Farbe Yellow
Einwegprodukt Reusable
Taillenform Non-elastic
Knöchelform Attached Socks, Überlappende Stiefel
Stulpenform Doppelt, Thumb Loop, Fingerschlaufen
Flaschenhalstyp Hooded
Hood Type Attached, Elastic Face
Verschlusstyp Zipper
Saumtyp Geschweißt, Geklebt
Sterilität Unsteril
Schutzartklasse CE-KATEGORIE III
Widerstand Antistatisch, Chemikalien, Partikel
Zertifizierungen/Konformität (Europa) CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Anzahl Taschen Keine
Zur Verwendung mit (Geräte) Opening and draining pumps valves or lines Filling, blending and charging of raw materials Contaminated Environments Chemical Handling Transferring liquids and solids Loading and unloading of proces equipment Raw material sample collection Concentration mixing and palletisation Hazmat response Assembly and disassembly of large pipes