Learn More
Amezinium methylsulfate, MedChemExpress
Artikelnummer. 16461986
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
100 mg
500 mg
1 g
Packungsgröße:
1 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16461986 500 mg -
16448046 100 mg -
16422029 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16461986 Lieferant Medchem Express Lieferanten-Nr. HYA0275
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Amezinium metilsulfate has multiple mechanisms, including stimulation of alpha and beta-1 receptors and inhibition ofnoradrenaline and tyramine uptake.
Amezinium metilsulfate has multiple mechanisms, including stimulation of alpha and beta-1 receptors and inhibition ofnoradrenaline and tyramine uptake.
Spezifikation
Chemischer Name oder Material Amezinium methylsulfate
Physikalische Form Solid
CAS 30578-37-1
Farbe White
Menge 500 mg
Hinweise zur Reinheitsqualität Research
Summenformel C12H15N3O5S
Zur Verwendung mit (Anwendung) COVID-19-immunoregulation
Synonym Amezinium metilsulfate Lu-1631
Löslichkeitsinformationen H2O : 50 mg/mL (159.58 mM; Need ultrasonic) ∣DMSO : ≥ 34 mg/mL (108.51 mM)
SMILES COC1=CC(N)=CN=[N+]1C2=CC=CC=C2.O=S(OC)([O-])=O
Molekulargewicht (g/mol) 313.33
Formelmasse 313.33
Reinheit (%) 99.51%
Güte Research