Learn More
A1CF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541849
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MESNHKSGDG LSGTQKEAAL RALVQRTGYS LVQENGQRKY GGPPPGWDAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
A1CF Polyclonal antibody specifically detects A1CF in Human samples. It is validated for Western Blot.Spezifikation
A1CF | |
Polyclonal | |
Unconjugated | |
A1CF | |
1810073H04Rik; A1cf; A1cft; Acf; ACF64; ACF65; apo-B RNA editing protein; Apobec-1; APOBEC1 complementation factor; apobec-1 complementation factor; apobec-1 complementation factor (ACF) (ASP); Apobec-1 complementation factor APOBEC-1 stimulating protein; Apobec-1 complementation factor, APOBEC-1 stimulating protein; APOBEC1 complementation factort; APOBEC-1 stimulating protein; APOBEC1CF; APOBEC1-stimulating protein; ASP; MGC163391; RP11-564C4.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
29974 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NQ94 | |
A1CF | |
Synthetic peptide directed towards the N-terminal of human A1CF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |