missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA9 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518953
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ABCA9 Polyclonal antibody specifically detects ABCA9 in Human samples. It is validated for Immunohistochemistry.Spezifikation
ABCA9 | |
Polyclonal | |
Unconjugated | |
ABCA9 | |
ABCA9; ATP binding cassette subfamily A member 9; ATP-binding cassette A9; ATP-binding cassette sub-family A member 9; ATP-binding cassette transporter sub-family A member 9; ATP-binding cassette, subfamily A (ABC1), member 9; ATP-binding cassette, sub-family A (ABC1), member 9; D630040K07Rik; DKFZp686F2450; EST640918; MGC75415 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10350 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8IUA7 | |
ABCA9 | |
Synthetic peptide sequence (QRVVQELEMENIQD) corresponding to the internal amino acids of ABCA9 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |