Learn More
ABCF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529955
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T. Predicted reactivity: Human (99%), Pig (89%), Rhesus Monkey (97%), Chimpanzee (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ABCF1 Polyclonal antibody specifically detects ABCF1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ABCF1 | |
Polyclonal | |
Unconjugated | |
Abcf1 | |
ABC27; ABC50; ABCF1; ATP binding cassette subfamily F member 1; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette sub-family F member 1; LOW QUALITY PROTEIN: ATP-binding cassette sub-family F member 1; ATP-binding cassette, subfamily F (GCN20), member 1; ATP-binding cassette, sub-family F (GCN20), member 1; ATP-binding cassette, sub-family F, member 1; AU041969; D17Wsu166e; GCN20; TNF-alpha stimulated ABC protein; TNFalpha-inducible ATP-binding protein; TNF-alpha-stimulated ABC protein; wu:fb79c06; wu:fc39a05; wu:fj94a08; zgc:85667 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NE71 | |
Abcf1 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 192 of Human ABCF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |