missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539006
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Acyl-CoA thioesterases (ACOTs) are a group of enzymes that catalyze the hydrolysis of acyl-CoA to form coenzyme A (CoA) and a free fatty acid. Through their catalytic activity, ACOTs are able to regulate the level of fatty acids and acyl-CoAs within the cell. ACOT1 (acyl-CoA thioesterase 1, also known as CTE1) and ACOT2 (acyl-CoA thioesterase 2, also known as PTE2) are members of the ACOT family and exhibit different cellular localization, with ACOT1 existing as a monomer in the cytoplasm and ACOT2 localized primarily to mitochondria. Characteristic of most ACOT proteins, ACOT1 and ACOT2 catalyze the conversion of Palmitoyl-CoA and water to free CoA and palmitate, a reaction that is important for the regulation of intercellular fatty acid levels. ACOT2 is expressed as multiple alternatively spliced isoforms and, like ACOT1, is encoded by a gene which maps to human chromosome 14.Spezifikation
ACOT2 | |
Polyclonal | |
Unconjugated | |
ACOT2 | |
ACOT2, Acyl CoA thioesterase 2, Acyl coenzyme A thioester hydrolase 2a, Acyl coenzyme A thioesterase 2, mitochondrial, CTE Ia, CTE1A, Long chain acyl CoA thioesterase 2, Mitochondrial acyl CoA thioesterase 1, Mitochondrial acyl CoA thioesterase 2, MTE1, Peroxisomal long chain acyl coA, PTE2, PTE2A, ZAP128 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10965 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49753 | |
ACOT2 | |
A synthetic peptide derived from the internal region of human ACOT2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |