Learn More
ACOT9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530476
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, U87-MG. Predicted reactivity: Mouse (81%), Rat (81%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acyl-CoA thioesterases (ACOTs) are a group of enzymes that catalyze the hydrolysis of acyl-CoA to form coenzyme A (CoA) and a free fatty acid. Through their catalytic activity, ACOTs are able to regulate the level of fatty acids and acyl-CoAs within the cell. ACOT9 (acyl-CoA thioesterase 9), also known as ACATE2, MT-ACT48 (mitochondrial acyl-CoA thioesterase of 48 kDa) or CGI-16, is a 406 amino acid member of the acyl-CoA hydrolase protein family. ACOT9 contains a C-terminal 80-amino acid domain that is conserved from mouse to human, suggesting that the C-terminus may confer the catalytic activity of ACOT9. The gene encoding ACOT9 is located on chromosome X and the expressed ACOT9 protein is localized to the mitochondrion.Spezifikation
ACOT9 | |
Polyclonal | |
Unconjugated | |
ACOT9 | |
0610041P13Rik; Acate2; ACOT9; Acyl coenzyme A thioester hydrolase 2; acyl-CoA thioester hydrolase 9; Acyl-CoA thioesterase 9; acyl-Coenzyme A thioesterase 2, mitochondrial; acyl-coenzyme A thioesterase 9, mitochondrial; C76421; CGI-16; Mitochondrial 48 kDa acyl-CoA thioester hydrolase 1; mitochondrial acyl-CoA thioesterase; MTACT48; MT-ACT48; mt-ACT48.1; MTE-2; p48; Protein U8; U8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23597 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.84 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y305 | |
ACOT9 | |
Recombinant fragment corresponding to a region within amino acids 1 and 234 of Human ACOT9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |