Learn More
ORM1 Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marke: Invitrogen PA19530
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-9530 detects Alpha1 Acid Glycoprotein in human samples. PA1-9530 has been successfully used in Western blot and ELISA procedures. By Western blot, PA1-9530 detects a ∽22 kDa protein representing Alpha1 Acid Glycoprotein. PA1-9530 immunogen is a mixture of synthetic peptides corresponding to residues 149-160 and 190-201 of human alpha-1 acid glycoprotein protein.
ORM1 Polyclonal antibody specifically detects ORM1 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
ORM1 | |
Polyclonal | |
Unconjugated | |
ORM1 | |
AGP; AGP 1; Agp1; Agp-1; Agp-2; AGP-A; Agpa1; alpha-1 acid glycoprotein; alpha-1-acid glycoprotein; Alpha-1-acid glycoprotein 1; alpha-1-acid glycoprotein-2; bM340H1.2 (orosomucoid 1); epididymis secretory sperm binding protein Li 153w; HEL-S-153w; OMD; OMD 1; Orm; Orm1; Orm-1; Orosomucoid; orosomucoid 1; orosomucoid-1 | |
Chicken | |
IgY fraction | |
RUO | |
5004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P02763 | |
ORM1 | |
Mixture of synthetic peptides corresponding to residues 149-160 and 190-201 of human alpha-1 acid glycoprotein protein. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgY |