Learn More
AP2 alpha Rabbit anti-Human, Mouse, Ovine, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA125327
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-25327 detects AP2 alpha from human samples. PA1-25327 is expected to cross react with mouse (94% conserved), rat (94% conserved), chicken (88% conserved), and ovine (94% conserved) due to sequence homology. PA1-25327 has been successfully used in Western blot and immunofluorescence procedures. The PA1-25327 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues S(420) H T D N N A K S S D K E E K H R K(437) of human AP2 alpha. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The protein encoded by this gene is a transcription factor that binds the consensus sequence 5′-GCCNNNGGC-3′. The encoded protein functions as either a homodimer or as a heterodimer with similar family members. This protein activates the transcription of some genes while inhibiting the transcription of others. Defects in this gene are a cause of branchiooculofacial syndrome (BOFS). Three transcript variants encoding different isoforms have been found for this gene.Spezifikation
AP2 alpha | |
Polyclonal | |
Unconjugated | |
P05549, P34056, Q9N0N3, P58197 | |
TFAP2A | |
Synthetic peptide, conjugated to KLH, corresponding to residues S(420) H T D N N A K S S D K E E K H R K(437) of human AP2 alpha. | |
100 μL | |
Primary | |
Human, Mouse, Ovine, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.9 mg/mL | |
TFAP2A | |
AP-2, AP-2alpha, AP2TF, BOFS, FLJ51761, TFAP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21418, 306862, 443109, 7020 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |