Learn More
APAAP Complex Monoclonal Antibody (WR31), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA180405
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-80405 detects APAAP Complex: an alkaline phosphatase linked to a monoclonal anti-alkaline phosphatase antibody. APAAP complexes are designed for use as detection reagents in immunohistochemistry procedures. They are used in combination with a suitable bridging antibody and appropriate visualization system.
APAAP Complex Monoclonal antibody specifically detects APAAP Complex in Chemical samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Spezifikation
APAAP Complex | |
Monoclonal | |
Unconjugated | |
Alkaline Phosphatase Complex | |
Bovine alkaline phosphatase. | |
1 mL | |
Primary | |
Antibody | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
WR31 | |
antibody with cell culture media and 0.09% sodium azide | |
Mouse | |
purified | |
RUO | |
4° C, do not freeze | |
Liquid | |
Chemical |