missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAALC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA513554
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
BAALC Polyclonal antibody specifically detects BAALC in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BAALC | |
Polyclonal | |
Unconjugated | |
BAALC | |
2810457D07Rik; Baalc; Brain and acute leukemia cytoplasmic protein; brain and acute leukemia, cytoplasmic; FLJ12015; HGNC:14333 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
79870 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q8WXS3 | |
BAALC | |
KLH conjugated synthetic peptide between 41-68 amino acids from the N-terminal region of human BAALC | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |