Learn More
BAG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529585
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (81%), Pig (85%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BAG1 Polyclonal antibody specifically detects BAG1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BAG1 | |
Polyclonal | |
Unconjugated | |
Bag1 | |
BAG family molecular chaperone regulator 1; Bag1; BAG-1; BCL2 associated athanogene 1; Bcl-2 associating athanogene-1 protein; BCL2-associated athanogene; bcl-2-associated athanogene 1; BCL2-associated athanogene 1; Bcl-2-binding protein; glucocortoid receptor-associated protein RAP46; HAP; Rap46; receptor-associated protein, 46-KD; RP11-326F20.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
573 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q99933 | |
Bag1 | |
Recombinant fragment corresponding to a region within amino acids 103 and 325 of Human Bag1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |