missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518481
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
GRK2 Polyclonal antibody specifically detects GRK2 in Human samples. It is validated for Western Blot.Spezifikation
GRK2 | |
Polyclonal | |
Unconjugated | |
GRK2 | |
ADRBK1; Adrbk-1; adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; BARK; BARK1; Bark-1; beta ARK; Beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; betaARK1; BETA-ARK1; beta-ARK-1; EC 2.7.1.126; G protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2; G-protein-coupled receptor kinase 2; grk 2; Grk2; GRK-2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
156 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P25098 | |
GRK2 | |
Synthetic peptide sequence (RDAYREAQQLVQR) corresponding to the internal amino acids of ADRBK1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |